Z277789
HGS Researcher model set
Inorganic chemistry-D
About This Item
Código UNSPSC:
60104506
NACRES:
NB.17
Produtos recomendados
Características
no. of atoms 116
no. of bonds 180
fabricante/nome comercial
HGS 1006
Descrição geral
Contain a variety of polyhedrons and bonds to meet the broad demand of chemical research. Scale: 1Å = 2.5 cm.
Researcher organic set is suited for building fundamental organic compounds, polymers, steroids, and some simple complex salts.
Researcher inorganic set is suited for building complicated tetra-, hexa-, and octa-coordinated complex compounds and chelate compounds. The fundamental structure of ionic crystals such as rock salt, cesium chloride, etc. can be constructed without supplementary parts.
Crystal structure set permits assembly of nearly all the fundamental crystal structures in inorganic chemistry and crystallography, as well as inorganic giant molecules.
Researcher organic set is suited for building fundamental organic compounds, polymers, steroids, and some simple complex salts.
Researcher inorganic set is suited for building complicated tetra-, hexa-, and octa-coordinated complex compounds and chelate compounds. The fundamental structure of ionic crystals such as rock salt, cesium chloride, etc. can be constructed without supplementary parts.
Crystal structure set permits assembly of nearly all the fundamental crystal structures in inorganic chemistry and crystallography, as well as inorganic giant molecules.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z277789-1EA | 4061833578506 |