Z277703
HGS Polyhedron molecular model
student set, fundamental organic chemistry
About This Item
Código UNSPSC:
41100000
NACRES:
NB.17
Produtos recomendados
descrição
scale: 1 Å = 2.5 cm
Características
no. of atoms 52
no. of bonds 62
fabricante/nome comercial
HGS 1000
Categorias relacionadas
Descrição geral
The use of 14, 20, and 26-sided polyhedrons as atomic nuclei makes these models unique. The various bond angles and distances needed for the models can be precisely represented, and the symmetrical characteristics of the structures can be observed.
- Color-coded polyhedrons to indicate different elements
- Flexible polyacetal connecting bonds permit easy assembly of complicated molecules and models of strained compounds and distorted crystals
- Å scale provided with sets allows measurement of bond distances
- Bond puller included for easy disassemble
- Two types of orbital plates for p atomic orbitals and π molecular orbitals (some sets only)
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z277703-6EA | 4061833569009 |
Z277703-24X1EA | |
Z277703-1EA | 4061833308981 |