Z258733
Ace vacuum-sublimation apparatus
Repl. CAPFE O-ring for Z258717
About This Item
Código UNSPSC:
41121800
NACRES:
NB.76
Produtos recomendados
fabricante/nome comercial
Ace Glass 7855881
Categorias relacionadas
Descrição geral
The Ace vacuum-sublimation apparatus features a greaseless O-ring sealed flange with a quick-release clamp. The top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. The complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z258733-1EA | 4061832349077 |