Key Documents
Z189960
ACE heavy-wall powder funnels
Angled, top diam. 75 mm
About This Item
Código UNSPSC:
41121809
NACRES:
NB.43
Junta
joint ST/NS24/40
fabricante/nome comercial
Ace Glass 723806
diâmetro do topo
75 mm
Categorias relacionadas
Descrição geral
For pouring powders or liquids into ground-joint containers.