Key Documents
Z131288
Sigma-Aldrich
Nalgene® volumetric flask
volume 500 mL, accuracy: 0.40 mL, translucent polypropylene
Sinônimo(s):
nalgene, plastic flask, plastic flasks
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Materiais
long
translucent polypropylene
Junta
joint threaded neck24mm
esterilidade
non-sterile
Características
accuracy class B (ASTM E288 and ISO Standard 384)
closure type Screw cap
embalagem
pack of 1
fabricante/nome comercial
Nalgene® 4000-0500
faixa de medição de volume
500 mL, accuracy: 0.40 mL
capacidade
500 mL
Descrição geral
Gravimetrically calibrated TC/TD at 20°C individually, then marked with a permanent color graduation line. Meet Class B accuracy requirements of ASTM E288 and ISO Standard 384. No meniscus to confuse readings. Oval marking spot. Linerless PP screw closure. Leakproof and autoclavable.
Informações legais
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries