WHA10410380
Whatman® regenerated cellulose membrane filters
RC58 sheets, pore size 0.2 μm, L × W 300 mm × 600 mm
Sinônimo(s):
Z750581
About This Item
Código UNSPSC:
40161507
NACRES:
NB.24
Produtos recomendados
embalagem
pk of 5
fabricante/nome comercial
Whatman 10410380
Whatman Article No. 28415294 (US reference)
C × L
300 mm × 600 mm
tamanho de poro
0.2 μm
Categorias relacionadas
Descrição geral
Cytiva′s Whatman regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
Versatile regenerated cellulose syringe filter membranes
Syringe filters require high-quality microporous membrane filters that minimally impact sample analysis with extractable impurities. Regenerated cellulose membrane filters from Cytiva′s business have broad solvent compatibility, and they contribute very low extractable material in a wide variety of sample solvents. Thus, they are appropriate for sample preparation in many applications and as a standalone or syringe filter membrane.
Versatile regenerated cellulose syringe filter membranes
Syringe filters require high-quality microporous membrane filters that minimally impact sample analysis with extractable impurities. Regenerated cellulose membrane filters from Cytiva′s business have broad solvent compatibility, and they contribute very low extractable material in a wide variety of sample solvents. Thus, they are appropriate for sample preparation in many applications and as a standalone or syringe filter membrane.
Características e benefícios
- Spontaneous wetting with very good wet strength
- Chemical resistance suitable for a wide range of aqueous and organic media
- Mechanically stable with low protein binding
- Low extractable levels to minimize sample contamination
- Can be sterilized by all common methods
Outras notas
Field of Use : Any microbiological or biotechnological application, including, but not limited to application of membranes for diagnostic, medical, biological, pharmaceutical, consumer product and food and beverage purposes.
Informações legais
Nitrocellulose Membranes (FFHP, Immunopore and BA membranes)
Sold under license to DE10102744 and foreign equivalents thereof
Sold under license to DE10102744 and foreign equivalents thereof
Whatman is a registered trademark of Cytiva
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WHA10410380 | 4061834015208 |