PL800GB-9-10
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 9 (for 10 in. ports)
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
Produtos recomendados
Materiais
butyl rubber
tamanho
9 (for 10 in. ports)
Características
reusable
embalagem
pack of 1 pair
fabricante/nome comercial
Plas-Labs 800GB-9-10
Descrição geral
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Escolha uma das versões mais recentes:
Certificados de análise (COA)