DWK92812G-500
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with glass stopper
glass flask, capacity (500 mL), class A
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Produtos recomendados
Materiais
glass flask
glass stopper
Características
class A
embalagem
case of 6 ea
fabricante/nome comercial
Kimble® 92812G-500
capacidade
(500 mL)
Descrição geral
Class A Heavy Duty Wide Mouth Volumetric Flasks with Glass Stopper
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)