Documentos Principais
DWK227494-02G
Wheaton® Bod Bottle
capacity 60 mL, bottle diam. × H 43 mm × 115 mm, pennyhead glass cap (stopper)
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Materiais
pennyhead glass cap (stopper)
embalagem
pkg of 36 ea
fabricante/nome comercial
WHEATON® 227494-02G
diâmetro do frasco × A
43 mm × 115 mm
capacidade
60 mL
Descrição geral
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Características e benefícios
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Informações legais
WHEATON is a registered trademark of Wheaton Industries, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)