BR4600231
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 5 mL, with recirculation valve
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Características
DE-M marking
with recirculation valve
fabricante/nome comercial
BRAND 4600231
volume
5 mL
conector
(GL 28/S28)
GL 32
GL 38
S 40
Categorias relacionadas
Descrição geral
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Características e benefícios
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Outras notas
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Conteúdo relacionado
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.
Global Trade Item Number
SKU | GTIN |
---|---|
BR4600231-1EA | 4061831865783 |