BR41654
BRAND® graduated cylinder low form embossed scale
volume 500 mL, polypropylene
Sinônimo(s):
Graduated Cylinders, Plastics
About This Item
Código UNSPSC:
41121805
NACRES:
NB.43
Produtos recomendados
Materiais
polypropylene
fabricante/nome comercial
BRAND 41654
volume
500 mL
Categorias relacionadas
Descrição geral
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR41654-5EA | 4061831865523 |