Principaux documents
EP5490037006
Rotor for Eppendorf® Concentrator Plus
F-40-18-19, non aerosol tight, to hold, 18 x 10 mL flat-bottom tubes
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 18 x 10 mL flat-bottom tubes
to hold 18 x 19 x 66 mm
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490037006 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Description générale
Selection of rotors for the Eppendorf Concentrator Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG