Principaux documents
EP5490040007
Rotor for Eppendorf® Concentrator Plus
F-40-36-12, non aerosol tight, to hold, 36 x 1.5 mL flat-bottom tubes
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 36 x 1.5 mL flat-bottom tubes
to hold 36 x 12 x 32 mm
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490040007 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Description générale
Selection of rotors for the Eppendorf Concentrator Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG