Key Documents
A8349
Spectrafuge 7M, 16M and Minifuge Adapters
0.2 mL PCR tubes
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41103900
Nomenclature NACRES :
NB.22
Fabricant/nom de marque
Labnet
Technique(s)
PCR: suitable
cryopreservation: suitable
Description générale
Package Qty: Adapters supplied as a set of 6
Informations légales
Spectrafuge is a trademark of Labnet Intl., Inc.