Merck
Z765562
Tube rack for Benchmixer XL multi-tube vortexer
Holds 50 x 12 mm tubes
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.77
Leistungsmerkmale
Holds 50 x 12 mm tubes
Verpackung
pkg of 1 ea
Hersteller/Markenname
Benchmark Scientific BV1010-12
Allgemeine Beschreibung
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables use of 50 x 12 mm tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. Ideal for the QuEChERS pesticide analysis method.
Rechtliche Hinweise
Benchmixer is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)