Merck
Z741755
Manifold
3-Place
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
Materialien
polysulfone
Leistungsmerkmale
autoclavable
Includes necessary tubing – 1/8” i.d. silicone
Hersteller/Markenname
IBI ACCFL0006
Allgemeine Beschreibung
The manifold allows for the connection and simultaneous delivery of media to flow cells and for consolidation of effluent from cells to the waste bottle. Comes with 1/8” i.d. silicone tubing attached and is autoclavable and re-usable.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)