Merck
Z659320
Bandelin SONOPULS ultrasonic homogenizer accessories
flow-through vessel DG 3 for HD 2070 and HD 2200 homogenizers
Synonym(e):
sonification vessel
About This Item
UNSPSC-Code:
41101520
NACRES:
NB.74
Materialien
glass
Verpackung
pack of 1 ea
Hersteller/Markenname
Bandelin 538
Verfügbarkeit
available only in EU
Volumen × ID × H
15 mL × 20 mm × 65 mm
Allgemeine Beschreibung
Flow vessel DG with cooling jacket, for continuous sonication of larger quantities in the flow. The cooling jacket allows temperature control using liquid coolants during sonication via existing hose connections.
Leistungsmerkmale und Vorteile
  • Simultaneous sound and cooling
  • Optimal process observation
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)