Merck
Z640174
Eppendorf® epT.I.P.S. Biopur®, singles
volume 2-200 μL
Synonym(e):
pipetting tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
colorless
Sterilität
sterile
Verpackung
pack of 100 (individually wrapped)
Hersteller/Markenname
Eppendorf® 0030 010.035
Eppendorf® 022491148
Messbereich Volumen
2-200 μL
Länge
53 mm
Maximales Volumen
200 μL
Allgemeine Beschreibung
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Rechtliche Hinweise
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG