Merck
Z601578
Hettich® centrifuge adapters
for ROTINA 38/38R and ROTINA 420/420R Centrifuges, Adapter for 9-15 mL tubes (14-17x100 mm)
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.22
Leistungsmerkmale
Holds 9 mL tubes
Verpackung
pack of 4 ea
Hersteller/Markenname
Hettich 1329
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 1490, 1495, 1481 (buckets)
Allgemeine Beschreibung
  • compatible with bucket/carriers 1381 (Z601543), 1390 and 1430
  • compatible with blood/urine 9-10 mL collection tubes (15-16x92-102 mm)
Rechtliche Hinweise
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG