Merck
Z380326
Jars Nalgene® style 2117
capacity 500 mL
Synonym(e):
laboratory jars, nalgene jars
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Verbindung
Joints threaded neck (120)
Beschreibung
Autoclavable with closure completely unscrewed
Leistungsmerkmale
with cap
Verpackung
pkg of 4
Hersteller/Markenname
Nalgene® 2117-0500
Flaschendurchm. × H
112 mm × 88 mm
Kapazität
500 mL
Allgemeine Beschreibung
Transparent, with good chemical resistance.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)