Merck
TMO362150-5240
Nalgene® red caps
polypropylene cap, diam. × thread 27 mm × 24-145 mm, case of 1000 ea
Synonym(e):
bottle cap, bottle closure
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
polypropylene cap
red cap
Verpackung
case of 1000 ea
Hersteller/Markenname
Nalgene 362150-5240
diam. × thread
27 mm × 24-145 mm
Allgemeine Beschreibung
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries