TMO362150-1200
Nalgene® white caps
polypropylene, diam. × thread 22 mm × 20-415 mm, case of 2000 ea
Synonym(e):
leakproof closure, threaded cap
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene
white
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 362150-1200
Durchm. × Gewinde
22 mm × 20-415 mm
Verwandte Kategorien
Allgemeine Beschreibung
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)