OH30253073
Ohaus® Scout® Portable Balance
Model SKX622, weighing capacity 620 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
Empfohlene Produkte
Materialien
metal weighing plate
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30253073
Messbereich Wiegekapazität
620 g, resolution: 0.01 g
Waagschalendurchm.
12 cm
Allgemeine Beschreibung
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Anwendung
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Leistungsmerkmale und Vorteile
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
OH30253073-1EA | 4065266747249 |