Merck
MTCR1081A
MTC Bio BetterBox Storage Box With Hinged Lid
holds 81 x 1.5/2.0 mL tubes, assorted colors
Synonym(e):
autoclable lab storage, cryo storage, stacking western blot box
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.12
Materialien
assorted colors
polypropylene
Leistungsmerkmale
holds 81 x 1.5/2.0 mL tubes
Verpackung
pack of 5 ea
Hersteller/Markenname
MTC Bio R1081-A
L × B × H
160 mm × 150 mm × 54 mm
Allgemeine Beschreibung
BetterBox Storage Box with hinged lid, 81 x 1.5mL to 2.0mL Microtubes pack of 5, assorted colors
Autoclavable virgin polypropylene storage box exclusively designed to hold 81×1.5mL or 2.0mL microtubes neatly and securely in nine rows of nine tubes (snap-cap or screw-cap tubes fit equally well). The lid cannot become separated due to the sturdy hinge and locking clasp. A frosted marking spot is molded into the front edge and the top surface. Available in a rainbow pack of five colors with clear lids and bases, the actual tube rack (colored middle layer) can be snapped out, allowing the base to be easily filled with crushed ice or liquid. They are Freezable to -80° C. The special corner feet are designed to mate with the four grooves in the top of the lid making them interlock and exceptionally easy to stack and transport.
Leistungsmerkmale und Vorteile
  • Organized and secure storage
  • 81-place for 1.5mL microtubes
  • Black, alphanumeric identification
  • Removeable insert – accepts crushed ice
Rechtliche Hinweise
BetterBox is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)