Merck
EPAG-21-00
Utility Tray
for Eppendorf® Brunswick Innova® 2350 shaker, with non-skid rubber surface
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.73
Leistungsmerkmale
with non-skid rubber surface
Hersteller/Markenname
Eppendorf® AG-21-00 (Global)
Allgemeine Beschreibung
Platform for Eppendorf New Brunswick Innova 2000 Platform Shaker to hold 10 mL Erlenmeyer flasks in dedicated aluminum clips.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)