Merck
EP5910710003
Spare Sealings, for Eppendorf® S-4x400 Rotor
for aerosol-tight caps, pack of 4 ea, for Centrifuge 5910 R
About This Item
UNSPSC-Code:
41103913
NACRES:
NB.82
Leistungsmerkmale
for Centrifuge 5910 R
for aerosol-tight caps
Verpackung
pack of 4 ea
Hersteller/Markenname
Eppendorf® 5910710003
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 5910 R
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG