Merck
EP5810718007
Eppendorf® Swing-bucket rotor A-4-81
incl. 4 x 500 ml rectangular buckets
About This Item
UNSPSC-Code:
41103909
NACRES:
NB.82
Leistungsmerkmale
to hold 4 x 500 mL rectangular buckets
incl. 4 x 500 ml rectangular buckets
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 5810718007
Parameter
4,000 max. RCF (3,220 xg)
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models 5804/5804R/5810/5810R
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG