Merck
EP5363000233
Lid for Eppendorf® SmartBlock 0.5 – 2.0 mL
for use with Eppendorf ThermoMixer® F0.5/F1.5/F2.0/FP, plates, PCR 96, PCR 384, and DWP
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.77
Leistungsmerkmale
for use with Eppendorf ThermoMixer® F0.5/F1.5/F2.0/FP, plates, PCR 96, PCR 384, and DWP
Hersteller/Markenname
Eppendorf® 5363000233 (Global)
Methode(n)
PCR: suitable
Allgemeine Beschreibung
  • for Eppendorf ThermoMixer® F0.5/F1.5/F2.0 and FP
  • for Eppendorf® SmartBlock 0.5 – 2.0 mL, plates, PCR 96, PCR 384, and DWP
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf ThermoMixer is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)