Merck
EP5316000004
Eppendorf® SmartBlock Thermoblock
for ThermoMixer®C & ThermoStat C, for Eppendorf® Deepwell Plate 96/500 μL, lid
Synonym(e):
Staining, Staining Accessories
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.77
Leistungsmerkmale
lid
Holds 1 x deepwell plate
for Eppendorf® Deepwell Plate 96/500 μL
for ThermoMixer®C & ThermoStat C
Hersteller/Markenname
Eppendorf® 5316000004 (Global)
Parameter
100 °C max. temp.
1500 rpm speed
Allgemeine Beschreibung
Using different vessel sizes within your lab workflow? Flexibility has never been this easy. Eppendorf® offers a broad range of SmartBlocks for tubes from 0.2 mL to 50 mL as well as plates (MTP, DWP, and PCR plates 96 / 384) to use with Eppendorf ThermoMixer® C and Eppendorf® ThermoStat C. All SmartBlocks are equipped with the outstanding Eppendorf® QuickRelease system that makes the block exchange super fast and easy. Just press the lever on the front of the block and the block can be removed – no tools are needed, done in seconds.
Leistungsmerkmale und Vorteile
  • Optimized block design supports optimal temperature transfer within your sample to receive excellent results
  • Eppendorf® SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Every SmartBlock is individually inspected in production for temperature accuracy and is delivered incl. a serial-number specific certificate for documentation purpose
  • Insulated thermoblocks for ergonomic operation – you won′t burn your fingers (PhysioCare Concept)
  • Automatic block recognition for safe handling
  • Optimized block design guarantees maximum temperature transfer to the sample
  • Eppendorf® QuickRelease technology enables fast and simple block exchange – no need for tools
  • Limited to ThermoMixer C and ThermoStat C
Max. mixing frequency using DWP is 2,000 rpm (80°C and higher: limited to 1,000 rpm)
Maximum mixing frequency is 2,000 rpm when lid is used
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf ThermoMixer is a registered trademark of Eppendorf AG
Thermomixer is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)