Merck
EP022638955
Adapter for Eppendorf® A-2-MTP Rotor
holds 1 x 384-well PCR plate, pack of 2 ea
About This Item
UNSPSC-Code:
41103911
Leistungsmerkmale
holds 1 x 384-well PCR plate
Verpackung
pack of 2 ea
Hersteller/Markenname
Eppendorf® 022638955 (NA)
Methode(n)
PCR: suitable
centrifugation: suitable
Kompatibilität
for use with A-2-DWP-AT
for use with A-2-DWP
for use with A-4-62-MTP
for use with Rotors A-4-81 MTP/Flex
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG