Merck
EP0030113519
Sigma-Aldrich
Eppendorf® Conical Tubes
Forensic DNA Grade, volume 50 mL, colorless (with 2D SafeCode), pkg of 48 tubes (individually wrapped)
Synonym(e):
Eppendorf tubes, Eppendorfs
About This Item
UNSPSC-Code:
41103037
Materialien
colorless (with 2D SafeCode)
conical bottom
polypropylene
Beschreibung
PCR Tubes
Sterilität
sterile
Leistungsmerkmale
RNase and DNase free
Forensic DNA Grade
Verpackung
pkg of 48 ea (individually wrapped)
pkg of 48 tubes (individually wrapped)
Hersteller/Markenname
Eppendorf® 0030113519
Volumen
50 mL
Allgemeine Beschreibung
The prevention of contamination is one of the major challenges in forensic DNA analysis. In order to minimize the risk of contamination, forensic laboratories have established strict internal processes. Even more safety is expected from the implementation of the ISO 18385 standard. This standard specifies the demands on manufacturers of products which will be used in forensic DNA laboratories. Eppendorf Forensic DNA Grade products are in compliance with the sophisticated requirements of this standard.
The prevention of contamination is one of the major challenges in forensic DNA analysis. In order to minimize the risk of contamination, forensic laboratories have established strict internal processes. Even more safety is expected from the implementation of the ISO 18385 standard.This standard specifies the demands on manufacturers of products which will be used in forensic DNA laboratories. Eppendorf Forensic DNA Grade products are in compliance with the sophisticated requirements of this standard.
Leistungsmerkmale und Vorteile
  • 2-level coding for safe sample identification ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -86 °C to +121 °C
  • Manufactured from high-quality polypropylene for lighest sample purity
  • Off-the-shelf availability for comfortable handling
  • ID-specific tube information like lot numbers digitally available via (URL: https://www.eppendorf.com/safecode-data)[Eppendorf dataport] and website for comprehensive documentation
  • 2-level coding for safe sample identification ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -86 °C to +121 °C
  • Manufactured from high-quality polypropylene for lighest sample purity
  • Off-the-shelf availability for comfortable handling
  • ID-specific tube information like lot numbers digitally available via (URL: https://www.eppendorf.com/safecode-data)[Eppendorf dataport] and website for comprehensive documentation
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)