CLS6703
Corning® LSE™ Platform Rocker
Single PlatformSingle Platform, AC/DC input 115 V, AC/DC input 50/60 Hz, AC/DC input, Schuko plug
Synonym(e):
equipment replacement accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103811
Empfohlene Produkte
Aufnahme AC/DC
, UK plug
Leistungsmerkmale
Single Platform
Single Platform
Hersteller/Markenname
Corning 6703
Allgemeine Beschreibung
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
- Low profile and compact size
- 11-pound load capacity
- Non-slip rubber pads to prevent sample containers from moving during operation
- Double platform model available for increased capacity
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)