Merck
CLS6634
Corning® Step-R Repeating Pipettor Syringe Tips
volume (25.0 mL), sterile
Synonym(e):
pipet tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.02
Sterilität
sterile
Verpackung
pack of 25 ea
Hersteller/Markenname
Corning 6634
Volumen
(25.0 mL)
Allgemeine Beschreibung
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)