CLS6635
Corning® Step-R Repeating Pipettor Syringe Tips
volume (50.0 mL), sterile
Synonym(e):
pipet tips
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.02
Empfohlene Produkte
Sterilität
sterile
Verpackung
pack of 25 ea
Hersteller/Markenname
Corning 6635
Volumen
(50.0 mL)
Allgemeine Beschreibung
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)