CLS480105
Optional heads for Corning® LSE™ vortex mixers
optional head for 12 x 1.5/2.0 mL tubes (held horizontally), 1/cs
Synonym(e):
equipment replacement accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103814
NACRES:
NB.15
Empfohlene Produkte
Verpackung
case of 1
pack of 1
Hersteller/Markenname
Corning 480105
Eignung
suitable for (for use with Corning LSE vortex mixer)
Allgemeine Beschreibung
Optional heads for the Corning LSE vortex mixers: CLS6776, CLS6777 and CLS6775
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS480105-1EA | 4061825882352 |