Merck
CLS1500P2L
Corning® narrow mouth reagent bottle, reusable
capacity 2 L, polypropylene, with GL-63 PP screw cap
Synonym(e):
laboratory bottles, plastic bottles, reagent bottles
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
cylindrical bottle
narrow-mouth bottle
polypropylene
screw top cap
translucent bottle
Leistungsmerkmale
cap
Verpackung
pack of 6 ea
Hersteller/Markenname
Corning 1500P-2L
H × AD
204 mm × 108 mm
Kapazität
2 L
Fitting
45
Allgemeine Beschreibung
These bottles are manufactured from translucent polypropylene. They are supplied with PP screw top caps.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
CLS1500P2L-6EA