BMSW3130
Serial Printer for Accuris™ Series Dx and Tx Balances
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103900
NACRES:
NB.61
Empfohlene Produkte
Hersteller/Markenname
Benchmark Scientific W3130
Allgemeine Beschreibung
This portable thermal printer offers a small footprint, modern design, and is compatible with the Accuris Dx and Tx Analytical Balances. Connection is easy with the included cables, and weight values and other critical info can be printed quickly and clearly. The printer connects to AC power (100V to 240V) with an included power adapter, and also features an included rechargeable Li-Ion battery.
Leistungsmerkmale und Vorteile
Compact and convenient
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)