BMSIPS205040
Benchmark OS-40L Overhead Stirrer
AC/DC input 120 V
Synonym(e):
overhead electric stirrer, digital overhead stirrer
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103800
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific IPS2050-40
Parameter
30-2200 rpm speed
Verwandte Kategorien
Allgemeine Beschreibung
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Leistungsmerkmale und Vorteile
LCD control panelIncludes complete stand assemblyCompact and quietDigital accuracy and convenience
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)