Merck
BAF187450012
Bel-Art® Switch-Grid® Test Tube Rack
Holds 40 x 16-20 mm tubes, green grid
Synonym(e):
tube holder
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
green grid
polyoxymethylene
Leistungsmerkmale
autoclavable
Holds 40 x 16-20 mm tubes
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F18745-0012
L × B × H
31.8 cm × 11.1 cm × 6.7 cm
Allgemeine Beschreibung
1 RACK FRAME, 25 POSSIBLE TUBE SIZE COMBINATIONS
This innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased as needed.
Leistungsmerkmale und Vorteile
More compact and less expensive than purchasing multiple racks
Colored grid inserts correspond to a specific test tube size ranging from 10mm to 30mm in diameter
Three tier rack holds tubes securely in place
Rubber feet keep racks from sliding on benchtop, provide secure stacking while in storage
Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F).
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Switch-Grid is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)