Merck
T2629
Sigma-Aldrich
Natriumwolframat Dihydrat
suitable for preparation of protein-free filtrates according to Folin
Synonym(e):
Wolframsäure Natriumsalz Dihydrat
About This Item
Lineare Formel:
Na2WO4 · 2H2O
CAS-Nummer:
Molekulargewicht:
329.85
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352302
PubChem Substanz-ID:
NACRES:
NA.55
Form:
powder
Form
powder
Eignung der Reaktion
reagent type: catalyst
core: tungsten
pH-Wert
9-11.5 (20 °C, 100 g/L)
mp (Schmelzpunkt)
698 °C (lit.)
Eignung
suitable for preparation of protein-free filtrates according to Folin
SMILES String
O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
InChI
1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;
InChIKey
WPZFLQRLSGVIAA-UHFFFAOYSA-N
Piktogramme
Exclamation mark
Signalwort
Warning
H-Sätze
Gefahreneinstufungen
Acute Tox. 4 Oral
Lagerklassenschlüssel
13 - Non Combustible Solids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Shannon K D Leblanc et al.
Journal of bacteriology, 193(13), 3367-3375 (2011-04-26)
The bacterial cell envelope is the interface between a bacterium and its environment and is constantly exposed to environmental changes. The BaeSR two-component system regulates one of six envelope stress responses in Escherichia coli and is induced by spheroplasting, overexpression
M Milagros Gómez-Gómez et al.
Talanta, 84(4), 1011-1018 (2011-05-03)
It is known that oral administration of sodium tungstate preserves the pancreatic beta cell function in diabetic rats. Healthy and streptozotocin-induced diabetic rats were treated with sodium tungstate for one, three or six weeks, after which the species of W
Cynthia D Fastje et al.
Chemico-biological interactions, 196(3), 89-95 (2011-05-14)
The etiology of childhood leukemia is not known. Strong evidence indicates that precursor B-cell Acute Lymphoblastic Leukemia (Pre-B ALL) is a genetic disease originating in utero. Environmental exposures in two concurrent, childhood leukemia clusters have been profiled and compared with
C J Rodriguez-Hernandez et al.
FEBS letters, 587(10), 1579-1586 (2013-04-17)
Both radiotherapy and most effective chemotherapeutic agents induce different types of DNA damage. Here we show that tungstate modulates cell response to DNA damaging agents. Cells treated with tungstate were more sensitive to etoposide, phleomycin and ionizing radiation (IR), all
F Hanzu et al.
Diabetes, obesity & metabolism, 12(11), 1013-1018 (2010-10-01)
Considering the poor long-term success of current dietary and pharmacological interventions, we aimed to evaluate the potential effect of sodium tungstate in the treatment of grade I and II obesity (ClinicalTrials.gov identifier: NCT00555074). Prospective, randomized, placebo-controlled, double-blind, proof-of-concept study was