YY1412224
1-1/2 in. Tri-Clover spacer with 1/4 in. NPTF Drain
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161525
eCl@ss:
32031603
Empfohlene Produkte
Materialien
stainless steel (Series 300)
Qualitätsniveau
ID
3.8 cm (1.5 in.)
Kompatibilität
for use with Series 2000 Sanitary Cartridge Housings