Wichtige Dokumente
MIX0010LLIN
Millipore
Mobius® Mixer Liner
Bronze Mobius® 10L mixer liner
Synonym(e):
Mobius® 10 L mixer liner, Bronze
About This Item
UNSPSC-Code:
41103801
eCl@ss:
32010600
Qualitätsniveau
Produktlinie
Mobius®
Rechtliche Hinweise
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany