Merck
Z760323
Ohaus® MB-23 and MB-25 moisture analyzers
Model MB25, AC/DC input 110 V AC
Synonym(e):
moisture meters
About This Item
UNSPSC-Code:
41111502
Hersteller/Markenname
Ohaus® MB25 US
Messbereich Wiegekapazität
110 g
Parameter
50-160 °C temp. limit
W × H × depth
17 cm × 13 cm × 28 cm
Plattformdurchm.
90 mm
Allgemeine Beschreibung
The MB25 has a halogen lamp for fast, even heating.
The Moisture analyzers combine high quality construction and easy setup. The LCD display is backlit, and it has a standard RS232 port.
  • easy to clean design
  • heating chamber is low maintenance
  • compact footprint
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)