Z757004
Kern DBS moisture analyzer, for samples up to 60 g
AC/DC input 220 V AC, UK plug
Synonym(e):
moisture sensor, moisture tester
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Kern DBS 60-3 GB
Messbereich Wiegekapazität
60 g
L × B × H
202 mm × 336 mm × 157 mm
Allgemeine Beschreibung
- Halogen quartz glass heater
- Observation window over the sample
- Internal memory for automatic sequence of 10 drying programs
- Sample description for up to 99 samples
- Date and time display as standard
- 10 sample plates included
Ähnliches Produkt
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z757004-1EA |