Merck
Z742528
Block for digital dry bath and MyBlock
for 2 or 4 block models, Holds 12 x 5 mL centrifuge tubes (17mm diameter) OR microplate, skirted or non-skirted
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.22
Leistungsmerkmale
Holds 12 x 5 mL centrifuge tubes (17mm diameter) OR microplate, skirted or non-skirted
for 2 or 4 block models
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific BSWMT
Allgemeine Beschreibung
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Rechtliche Hinweise
MyBlock is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z742528-1EA4061826161654