Z742516
Block for digital dry bath and MyBlock™
Holds 24 x 1.5 mL tubes
Alle Fotos(1)
About This Item
UNSPSC-Code:
41102404
NACRES:
NB.77
Empfohlene Produkte
Leistungsmerkmale
Holds 24 x 1.5 mL tubes
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific BSW1500
Allgemeine Beschreibung
For use with Benchmark Digital Dry Bath
- Material: high grade aluminum
- Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Rechtliche Hinweise
MyBlock is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)