Merck
Z742355
2MAG Extension cord for bioMIXdrive
L 3 m
Synonym(e):
stirrer control unit cable
About This Item
UNSPSC-Code:
41116500
NACRES:
NB.72
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 46200
Parameter
-10-50 °C temp. limit (at 80% humidity)
Länge
3 m
Allgemeine Beschreibung
Extension cord for use with bioMixdrive stirring drive, 3m in length. Perm Operating Temperature -10°C up to +50°C (at 80% humidity).
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)