Z740943
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 10 mL, O.D. × L 15 mm × 125 mm, round bottom
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.11
Empfohlene Produkte
Materialien
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
Verpackung
pack of 72 ea
Hersteller/Markenname
Sigma-Aldrich
AD × L
15 mm × 125 mm
Kapazität
10 mL
Verwandte Kategorien
Allgemeine Beschreibung
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z740943-72EA | 4061833641538 |