Z740940
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 5 mL, O.D. × L 15 mm × 75 mm, round bottom
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.11
Empfohlene Produkte
Materialien
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
Verpackung
pack of 72 ea
Hersteller/Markenname
Sigma-Aldrich
AD × L
15 mm × 75 mm
Kapazität
5 mL
Verwandte Kategorien
Allgemeine Beschreibung
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z740940-72EA | 4061833641521 |