Z682209
Ace receiving flask adapter
joint: ST/NS 29/32, fits Büchi® models R-200/R-205
Synonym(e):
Ace receiving flask adapter
Empfohlene Produkte
Verbindung
joint: ST/NS29/32
Hersteller/Markenname
Büchi equivalent, 40615
Allgemeine Beschreibung
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)